Assay Details
Gene Symbol: Kctd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105768.1 | NP_001099238.1 | 4 - 5 | 566 | 80 | |
GenBank mRNA | BC168712.1 | - | 4 - 5 | 569 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
