Assay Details
Gene Symbol: Mnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105807.1 | NP_001099277.1 | 3 - 4 | 1101 | 77 | |
XM_008767959.1 | XP_008766181.1 | 3 - 4 | 11748 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
