Assay Details
Gene Symbol: Crebrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277157.1 | NP_001264086.1 | 2 - 3 | 247 | 59 | |
XM_008767581.1 | XP_008765803.1 | 1 - 2 | 467 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277157.1, XM_008767581.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
