Assay Details
Gene Symbol: Alox12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039377.1 | NP_001034466.1 | 9 - 10 | 1413 | 63 | |
GenBank mRNA | AY903231.1 | - | 9 - 10 | 1413 | 63 | |
AY903232.1 | - | 9 - 10 | 1413 | 63 | ||
AY903233.1 | - | 9 - 10 | 1413 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
