Assay Details
Gene Symbol: Afmid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111366.1 | NP_001104836.1 | 8 - 9 | 676 | 73 | |
XM_006247823.2 | XP_006247885.1 | 8 - 9 | 692 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
