Assay Details
Gene Symbol: Mrps23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108289.1 | NP_001101759.1 | 3 - 4 | 317 | 95 | |
XM_008768075.1 | XP_008766297.1 | 3 - 4 | 508 | 95 | ||
GenBank mRNA | FQ216939.1 | - | 3 - 4 | 348 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
