Assay Details
Gene Symbol: Mrps23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108289.1 | NP_001101759.1 | 1 - 2 | 72 | 58 | |
GenBank mRNA | FQ213501.1 | - | 1 - 2 | 608 | 58 | |
FQ216939.1 | - | 1 - 2 | 103 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
