Assay Details
Gene Symbol: Pycr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105857.2 | NP_001099327.1 | 1 - 2 | 66 | 79 | |
XM_008768472.1 | XP_008766694.1 | 2 - 3 | 263 | 79 | ||
XM_008768473.1 | XP_008766695.1 | 2 - 3 | 197 | 79 | ||
XM_008768474.1 | XP_008766696.1 | 2 - 3 | 187 | 79 | ||
XM_008768475.1 | XP_008766697.1 | 2 - 3 | 263 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
