Assay Details
Gene Symbol: Shmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047842.1 | NP_001041307.1 | 11 - 12 | 1526 | 67 | |
XM_006246469.2 | XP_006246531.1 | 8 - 9 | 1309 | 67 | ||
GenBank mRNA | AY383687.1 | - | 11 - 12 | 1526 | 67 | |
BC099219.1 | - | 4 - 5 | 540 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
