Assay Details
Gene Symbol: Shmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047842.1 | NP_001041307.1 | 14 - 15 | 1872 | 92 | |
XM_006246469.2 | XP_006246531.1 | 11 - 12 | 1655 | 92 | ||
GenBank mRNA | AY383687.1 | - | 14 - 15 | 1872 | 92 | |
BC099219.1 | - | 7 - 8 | 886 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
glycine metabolic process
glycine biosynthetic process
L-serine metabolic process
L-serine catabolic process
purine nucleobase biosynthetic process
glycine biosynthetic process from serine
tetrahydrofolate interconversion
folic acid metabolic process
protein tetramerization
protein homotetramerization
glycine hydroxymethyltransferase activity
L-allo-threonine aldolase activity
amino acid binding
pyridoxal phosphate binding
identical protein binding
protein homodimerization activity
glycine metabolic process
glycine biosynthetic process
L-serine metabolic process
L-serine catabolic process
purine nucleobase biosynthetic process
glycine biosynthetic process from serine
tetrahydrofolate interconversion
folic acid metabolic process
protein tetramerization
protein homotetramerization
glycine hydroxymethyltransferase activity
L-allo-threonine aldolase activity
amino acid binding
pyridoxal phosphate binding
identical protein binding
protein homodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P132367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162328 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132367 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132367 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P132367 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1055774 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P160813 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P508239 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14508 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 93245 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10743323 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743325 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743329 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743330 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743328 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743320 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743322 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743326 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743332 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743333 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743327 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743319 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743331 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743324 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743321 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10743318 | - | - |
Rat Genome 230 2.0 Array | 1374748_at | - | - |
Rat Genome 230 2.0 Array | 1373625_at | - | - |
Rat Genome 230 2.0 Array | 1373625_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374748_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374748_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373625_at | - | - |
Affymetrix HT Rat Focus Array | 1374748_at | - | - |
Affymetrix HT Rat Focus Array | 1373625_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000025872.rn.1 | TC1000002942.rn.1 | RTA TC |
Rat Expression Set 230 | 1373625_at | - | - |
Rat Expression Set 230 | 1373625_PM_at | - | - |
Rat Expression Set 230 | 1374748_PM_at | - | - |
Rat Expression Set 230 | 1374748_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743318 | - | - |
Rat Genome U34 Set | rc_AI011901_at | - | - |
Rat Genome U34 Set | rc_AI012348_at | - | - |
Rat Genome U34 Set | rc_AI011901_g_at | - | - |
Rat Genome U34 Set | rc_AA819027_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20930758 | - | - |
ABI Rat Genome Survey Microarray | 21771064 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356184 | - | - |
