Assay Details
Gene Symbol: Fbxw11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106993.1 | NP_001100463.1 | 7 - 8 | 857 | 101 | |
XM_006246092.2 | XP_006246154.1 | 8 - 9 | 1114 | 101 | ||
XM_006246093.2 | XP_006246155.1 | 8 - 9 | 1105 | 101 | ||
XM_006246094.2 | XP_006246156.1 | 7 - 8 | 1042 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
