Assay Details
Gene Symbol: Socs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247484.2 | XP_006247546.2 | 7 - 8 | 1730 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
