Assay Details
Gene Symbol: Pigq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007607.1 | NP_001007608.1 | 3 - 4 | 869 | 56 | |
XM_006245942.2 | XP_006246004.1 | 3 - 4 | 948 | 56 | ||
XM_008767559.1 | XP_008765781.1 | 3 - 4 | 888 | 56 | ||
XM_008767560.1 | XP_008765782.1 | 3 - 4 | 915 | 56 | ||
GenBank mRNA | BC081875.1 | - | 3 - 4 | 869 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
