Assay Details
Gene Symbol: Arhgdia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007005.1 | NP_001007006.1 | 4 - 5 | 430 | 69 | |
XM_006247897.2 | XP_006247959.1 | 4 - 5 | 564 | 69 | ||
XM_006247898.2 | XP_006247960.1 | 4 - 5 | 513 | 69 | ||
GenBank mRNA | BC083817.1 | - | 4 - 5 | 430 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
