Assay Details
Gene Symbol: Dnajc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213625.1 | NP_998790.1 | 7 - 8 | 749 | 124 | |
XM_006247311.2 | XP_006247373.1 | 8 - 9 | 1110 | 124 | ||
XM_006247312.2 | XP_006247374.1 | 7 - 8 | 830 | 124 | ||
XM_006247313.1 | XP_006247375.1 | 6 - 7 | 670 | 124 | ||
GenBank mRNA | AB178470.1 | - | 7 - 8 | 749 | 124 | |
BC093600.1 | - | 6 - 7 | 665 | 124 | ||
FQ212948.1 | - | 7 - 8 | 780 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
