Assay Details
Gene Symbol: Eda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257110.2 | XP_006257172.1 | 7 - 8 | 1124 | 70 | |
XM_006257112.2 | XP_006257174.1 | 7 - 8 | 1115 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
