Assay Details
Gene Symbol: Il2rg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080889.1 | NP_543165.1 | 2 - 3 | 285 | 69 | |
GenBank mRNA | AF410926.1 | - | 2 - 3 | 205 | 69 | |
BC079343.1 | - | 2 - 3 | 285 | 69 | ||
FQ232752.1 | - | 1 - 2 | 86 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
