Assay Details
Gene Symbol: Igf2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270598.1 | NP_001257527.1 | 5 - 6 | 480 | 84 | |
NM_001270599.1 | NP_001257528.1 | 5 - 6 | 480 | 84 | ||
GenBank mRNA | FQ222567.1 | - | 5 - 6 | 248 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
