Assay Details
Gene Symbol: Alg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142363.1 | NP_001135835.1 | 3 - 4 | 466 | 82 | |
NR_024535.1 | 2 - 3 | 366 | 82 | |||
XM_006248558.2 | XP_006248620.1 | 3 - 4 | 484 | 82 | ||
GenBank mRNA | BC088475.1 | - | 2 - 3 | 349 | 82 | |
CO401508.1 | - | 3 - 4 | 455 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
