Assay Details
Gene Symbol: Ndfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108390.2 | NP_001101860.2 | 7 - 8 | 1116 | 91 | |
XM_006252429.1 | XP_006252491.1 | 8 - 9 | 1061 | 91 | ||
GenBank mRNA | FM044778.1 | - | 2 - 3 | 165 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P040159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901629 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30183 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P040159 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P040159 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P040159 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1047638 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19157 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42526 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32103 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10781948 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781946 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781947 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781945 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781944 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781942 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781949 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781943 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10781941 | - | - |
Rat Genome 230 2.0 Array | 1389364_at | - | - |
Rat Genome 230 2.0 Array | 1373861_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389364_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373861_at | - | - |
Affymetrix HT Rat Focus Array | 1373861_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373861_at | - | - |
Affymetrix HT Rat Focus Array | 1389364_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389364_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1500006900.rn.1 | TC1500001032.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500006899.rn.1 | TC1500001032.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500006901.rn.1 | TC1500001032.rn.1 | RTA TC |
Rat Expression Set 230 | 1373861_at | - | - |
Rat Expression Set 230 | 1389364_at | - | - |
Rat Expression Set 230 | 1373861_PM_at | - | - |
Rat Expression Set 230 | 1389364_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781941 | - | - |
Rat Genome U34 Set | rc_AI176927_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21954390 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361407 | - | - |
