Assay Details
Gene Symbol: Tnfrsf10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108873.1 | NP_001102343.1 | 4 - 5 | 671 | 67 | |
GenBank mRNA | FJ515907.1 | - | 5 - 6 | 673 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
