Assay Details
Gene Symbol: Lcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012044.1 | NP_001012044.1 | 11 - 12 | 1299 | 113 | |
XM_006252318.2 | XP_006252380.1 | 11 - 12 | 1474 | 113 | ||
XM_006252319.2 | XP_006252381.1 | 13 - 14 | 1673 | 113 | ||
XM_006252320.2 | XP_006252382.1 | 13 - 14 | 1721 | 113 | ||
XM_006252322.2 | XP_006252384.1 | 11 - 12 | 2880 | 113 | ||
GenBank mRNA | BC083855.1 | - | 11 - 12 | 1299 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
