Assay Details
Gene Symbol: Clybl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100685.1 | NP_001094155.1 | 2 - 3 | 264 | 73 | |
GenBank mRNA | BC088243.1 | - | 2 - 3 | 244 | 73 | |
FM063772.1 | - | 2 - 3 | 264 | 73 | ||
FQ214276.1 | - | 1 - 2 | 49 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
