Assay Details
Gene Symbol: Clybl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100685.1 | NP_001094155.1 | 7 - 8 | 933 | 76 | |
XM_006252508.2 | XP_006252570.1 | 6 - 7 | 762 | 76 | ||
GenBank mRNA | BC088243.1 | - | 7 - 8 | 913 | 76 | |
FQ214276.1 | - | 6 - 7 | 718 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P029062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70712 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P029062 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P029062 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P029062 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1013163 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103082 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51900 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10610 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2753 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4005 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98452 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17118 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1299 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46410 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42688 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12245 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10782147 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782149 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782151 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782153 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782150 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782152 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782145 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782148 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782146 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10782144 | - | - |
Rat Genome 230 2.0 Array | 1374190_at | - | - |
Rat Genome 230 2.0 Array | 1374190_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374190_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374190_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1500007533.rn.1 | TC1500001200.rn.1 | RTA TC |
Rat Expression Set 230 | 1374190_PM_at | - | - |
Rat Expression Set 230 | 1374190_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782144 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21080301 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356518 | - | - |
