Assay Details
Gene Symbol: Mtrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751518.3 | XP_003751566.1 | 10 - 11 | 1637 | 63 | |
XM_006252367.2 | XP_006252429.1 | 10 - 11 | 1675 | 63 | ||
XM_006252368.2 | XP_006252430.1 | 9 - 10 | 1527 | 63 | ||
XM_008770927.1 | XP_008769149.1 | 10 - 11 | 1366 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
