Assay Details
Gene Symbol: Mtrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751518.3 | XP_003751566.1 | 2 - 3 | 408 | 127 | |
XM_006252367.2 | XP_006252429.1 | 2 - 3 | 446 | 127 | ||
XM_006252369.2 | XP_006252431.1 | 2 - 3 | 408 | 127 | ||
XM_008770927.1 | XP_008769149.1 | 2 - 3 | 137 | 127 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003751518.3, XM_006252367.2, XM_006252369.2, XM_008770927.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
