Assay Details
Gene Symbol: Mtrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751518.3 | XP_003751566.1 | 4 - 5 | 927 | 103 | |
XM_006252367.2 | XP_006252429.1 | 4 - 5 | 965 | 103 | ||
XM_006252368.2 | XP_006252430.1 | 3 - 4 | 817 | 103 | ||
XM_006252369.2 | XP_006252431.1 | 4 - 5 | 927 | 103 | ||
XM_008770927.1 | XP_008769149.1 | 4 - 5 | 656 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
