Assay Details
Gene Symbol: Eif2s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199380.1 | NP_955412.1 | 7 - 8 | 819 | 157 | |
GenBank mRNA | BC062402.1 | - | 7 - 8 | 819 | 157 | |
FQ215337.1 | - | 7 - 8 | 864 | 157 | ||
FQ215587.1 | - | 7 - 8 | 760 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
