Assay Details
Gene Symbol: Eif2s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199380.1 | NP_955412.1 | 8 - 9 | 906 | 70 | |
GenBank mRNA | BC062402.1 | - | 8 - 9 | 906 | 70 | |
FQ209519.1 | - | 6 - 7 | 583 | 70 | ||
FQ213305.1 | - | 8 - 9 | 938 | 70 | ||
FQ215337.1 | - | 8 - 9 | 951 | 70 | ||
FQ215587.1 | - | 8 - 9 | 847 | 70 | ||
FQ234002.1 | - | 8 - 9 | 935 | 70 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
