Assay Details
Gene Symbol: Sdcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025692.1 | NP_001020863.1 | 2 - 3 | 131 | 64 | |
XM_008762326.1 | XP_008760548.1 | 3 - 4 | 1112 | 64 | ||
GenBank mRNA | BC099095.1 | - | 2 - 3 | 131 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
