Assay Details
Gene Symbol: Sdcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025692.1 | NP_001020863.1 | 3 - 4 | 211 | 74 | |
XM_008762325.1 | XP_008760547.1 | 1 - 2 | 2187 | 74 | ||
XM_008762326.1 | XP_008760548.1 | 4 - 5 | 1192 | 74 | ||
GenBank mRNA | BC099095.1 | - | 3 - 4 | 211 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
