Assay Details
Gene Symbol: Sdcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025692.1 | NP_001020863.1 | 7 - 8 | 815 | 68 | |
XM_008762325.1 | XP_008760547.1 | 5 - 6 | 2791 | 68 | ||
XM_008762326.1 | XP_008760548.1 | 8 - 9 | 1796 | 68 | ||
GenBank mRNA | BC099095.1 | - | 7 - 8 | 815 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
