Assay Details
Gene Symbol: Kif3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106529.1 | NP_001099999.1 | 8 - 9 | 2292 | 119 | |
XM_006235287.2 | XP_006235349.1 | 8 - 9 | 2713 | 119 | ||
XM_006235288.1 | XP_006235350.1 | 8 - 9 | 2329 | 119 | ||
XM_006235289.2 | XP_006235351.1 | 8 - 9 | 2509 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P082113 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P529939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168317 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P529939 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P082113 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P082113 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P529939 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P082113 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P529939 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P715184 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P529939 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P529947 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10841032 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841027 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841026 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841030 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841028 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841031 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841029 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841023 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10841022 | - | - |
Rat Genome 230 2.0 Array | 1374428_at | - | - |
Rat Genome 230 2.0 Array | 1379963_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379963_at | - | - |
Rat Genome 230 2.0 Array | 1374428_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374428_at | - | - |
Affymetrix HT Rat Focus Array | 1379963_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374428_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379963_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300014855.rn.1 | TC0300002064.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300014854.rn.1 | TC0300002064.rn.1 | RTA TC |
Rat Expression Set 230 | 1374428_PM_at | - | - |
Rat Expression Set 230 | 1374428_at | - | - |
Rat Expression Set 230 | 1379963_PM_at | - | - |
Rat Expression Set 230 | 1379963_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841022 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21931925 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352633 | - | - |
