Assay Details
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031068.1 | NP_112330.1 | 10 - 11 | 1163 | 87 | |
GenBank mRNA | AB292042.1 | - | 10 - 11 | 1087 | 87 | |
AB292043.1 | - | 10 - 11 | 1076 | 87 | ||
AF307852.1 | - | 1 - 2 | 80 | 87 | ||
BC098014.1 | - | 10 - 11 | 1163 | 87 | ||
FQ212256.1 | - | 10 - 11 | 1365 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
