Assay Details
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031068.1 | NP_112330.1 | 4 - 5 | 421 | 150 | |
GenBank mRNA | AB292043.1 | - | 4 - 5 | 334 | 150 | |
BC098014.1 | - | 4 - 5 | 421 | 150 | ||
FQ212256.1 | - | 4 - 5 | 623 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
