Assay Details
Gene Symbol: Nr6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761807.1 | XP_008760029.1 | 7 - 8 | 1219 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
