Assay Details
Gene Symbol: Tnfaip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053382.1 | NP_445834.1 | 4 - 5 | 639 | 62 | |
GenBank mRNA | BC158666.1 | - | 4 - 5 | 639 | 62 | |
FQ228771.1 | - | 4 - 5 | 701 | 62 | ||
FQ228783.1 | - | 4 - 5 | 701 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
