Assay Details
Gene Symbol: Rnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109025.2 | NP_001102495.1 | 3 - 4 | 372 | 260 | |
GenBank mRNA | BC088341.1 | - | 3 - 4 | 348 | 260 | |
DY573537.1 | - | 3 - 4 | 372 | 260 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
