Assay Details
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198744.2 | NP_942039.1 | 2 - 3 | 314 | 70 | |
XM_006235754.2 | XP_006235816.1 | 2 - 3 | 313 | 70 | ||
XM_006235757.2 | XP_006235819.1 | 2 - 3 | 333 | 70 | ||
XM_006235759.2 | XP_006235821.1 | 2 - 3 | 306 | 70 | ||
XM_006235760.2 | XP_006235822.1 | 2 - 3 | 328 | 70 | ||
XM_006235761.2 | XP_006235823.1 | 2 - 3 | 311 | 70 | ||
XM_008762499.1 | XP_008760721.1 | 2 - 3 | 247 | 70 | ||
XM_008762500.1 | XP_008760722.1 | 2 - 3 | 245 | 70 | ||
GenBank mRNA | BC059167.1 | - | 2 - 3 | 315 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
