Assay Details
Gene Symbol: Cables2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235831.2 | XP_006235893.2 | 6 - 7 | 893 | 78 | |
XM_008762586.1 | XP_008760808.1 | 6 - 7 | 893 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
