Assay Details
Gene Symbol: Srms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011961.1 | NP_001011961.1 | 7 - 8 | 1425 | 90 | |
GenBank mRNA | BC090006.1 | - | 7 - 8 | 1425 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
