Assay Details
Gene Symbol: Slc35e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109107.2 | NP_001102577.2 | 5 - 6 | 1012 | 77 | |
GenBank mRNA | CB316326.1 | - | 3 - 4 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
