Assay Details
Gene Symbol: Cyp2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108499.1 | NP_001101969.1 | 4 - 5 | 706 | 94 | |
XM_006230081.2 | XP_006230143.1 | 4 - 5 | 1717 | 94 | ||
XM_006230084.2 | XP_006230146.1 | 4 - 5 | 1496 | 94 | ||
GenBank mRNA | BC168986.1 | - | 3 - 4 | 1124 | 94 | |
FQ219437.1 | - | 3 - 4 | 1020 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
