Assay Details
Gene Symbol: Ucp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019354.3 | NP_062227.2 | 5 - 6 | 909 | 74 | |
GenBank mRNA | AB005143.1 | - | 3 - 4 | 536 | 74 | |
AB006613.1 | - | 3 - 4 | 536 | 74 | ||
AB010743.1 | - | 5 - 6 | 880 | 74 | ||
AF039033.1 | - | 3 - 4 | 536 | 74 | ||
BC062230.1 | - | 5 - 6 | 881 | 74 | ||
FQ227097.1 | - | 5 - 6 | 910 | 74 | ||
FQ230356.1 | - | 1 - 2 | 195 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
