Assay Details
Gene Symbol: Ucp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019354.3 | NP_062227.2 | 7 - 8 | 1189 | 87 | |
GenBank mRNA | AB005143.1 | - | 5 - 6 | 816 | 87 | |
AB006613.1 | - | 5 - 6 | 816 | 87 | ||
AF039033.1 | - | 5 - 6 | 816 | 87 | ||
FQ227097.1 | - | 7 - 8 | 1190 | 87 | ||
FQ230356.1 | - | 3 - 4 | 475 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
