Assay Details
Gene Symbol: Rab30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229692.2 | XP_006229754.1 | 2 - 3 | 307 | 63 | |
XM_006229693.2 | XP_006229755.1 | 2 - 3 | 276 | 63 | ||
XM_008759647.1 | XP_008757869.1 | 1 - 2 | 3274 | 63 | ||
GenBank mRNA | FQ210359.1 | - | 1 - 2 | 183 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
