Assay Details
Gene Symbol: Cyb5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014244.1 | NP_001014266.1 | 3 - 4 | 319 | 75 | |
XM_008759774.1 | XP_008757996.1 | 1 - 2 | 1457 | 75 | ||
XM_008759775.1 | XP_008757997.1 | 2 - 3 | 1360 | 75 | ||
XM_008759776.1 | XP_008757998.1 | 1 - 2 | 1457 | 75 | ||
XM_008759777.1 | XP_008757999.1 | 1 - 2 | 1457 | 75 | ||
XM_008759778.1 | XP_008758000.1 | 2 - 3 | 775 | 75 | ||
XM_008759779.1 | XP_008758001.1 | 2 - 3 | 932 | 75 | ||
XM_008759781.1 | XP_008758003.1 | 3 - 4 | 256 | 75 | ||
XM_008759782.1 | XP_008758004.1 | 1 - 2 | 1457 | 75 | ||
GenBank mRNA | BC079235.1 | - | 3 - 4 | 319 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
