Assay Details
Gene Symbol: Cyb5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014244.1 | NP_001014266.1 | 7 - 8 | 712 | 98 | |
XM_008759777.1 | XP_008757999.1 | 5 - 6 | 1850 | 98 | ||
XM_008759782.1 | XP_008758004.1 | 5 - 6 | 1898 | 98 | ||
GenBank mRNA | BC079235.1 | - | 7 - 8 | 712 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
