Assay Details
Gene Symbol: Eif4g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017374.2 | NP_001017374.2 | 10 - 11 | 1204 | 69 | |
GenBank mRNA | BC099117.1 | - | 10 - 11 | 1091 | 69 |
Gene Symbol: LOC501116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765067.1 | XP_008763289.1 | 2 | 1026 | 69 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
