Assay Details
Gene Symbol: Eif4g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017374.2 | NP_001017374.2 | 16 - 17 | 1941 | 75 | |
GenBank mRNA | AB256044.1 | - | 2 - 3 | 75 | ||
AM258959.1 | - | 2 - 3 | 75 | |||
BC091330.1 | - | 2 - 3 | 252 | 75 | ||
BC099117.1 | - | 16 - 17 | 1828 | 75 |
Gene Symbol: LOC501116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765067.1 | XP_008763289.1 | 2 | 1763 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
